missing translation for 'onlineSavingsMsg'
Learn More

1,1'-(Azodicarbonyl)dipiperidine 98.0+%, TCI America™

Catalog No. A10515G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10515G 5 g
A105125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10515G Supplier TCI America Supplier No. A10515G

Chemical Identifiers

CAS 10465-81-3
Molecular Formula C12H20N4O2
Molecular Weight (g/mol) 252.32
MDL Number MFCD00010111
InChI Key OQJBFFCUFALWQL-UHFFFAOYSA-N
Synonym 1,1'-azodicarbonyl-dipiperidine, 1,1-azodicarbonyl dipiperidine, 1,1'-azodicarbonyl dipiperidine, addp, diazene-1,2-diylbis piperidin-1-ylmethanone, azodicarboxylic acid dipiperidine, ne-n-piperidine-1-carbonylimino piperidine-1-carboxamide, 1,1'-azodicarbonyl dipiperidine addp, piperidyl piperidylcarbonyl diazenyl ketone, e-diazene-1,2-diylbis piperidin-1-ylmethanone
PubChem CID 5702657
IUPAC Name N-[(piperidine-1-carbonyl)imino]piperidine-1-carboxamide
SMILES O=C(N=NC(=O)N1CCCCC1)N1CCCCC1

Specifications

Melting Point 136°C
Quantity 5 g
Formula Weight 252.32
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-(Azodicarbonyl)dipiperidine