accessibility menu, dialog, popup

UserName

ONNONN(E/Z)

CAS RN 10465-81-3

IUPAC Name: N-[(piperidine-1-carbonyl)imino]piperidine-1-carboxamide
Synonyms: N-(piperidine-1-carbonylimino)piperidine-1-carboxamide
Molecular Weight (g/mol): 252.32
Molecular Formula: C12H20N4O2
InChi Key: OQJBFFCUFALWQL-UHFFFAOYSA-N
SMILES: O=C(N=NC(=O)N1CCCCC1)N1CCCCC1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)