missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Adamantyl)ethylamine hydrochloride, Thermo Scientific™

$19.14 - $19.14

Chemical Identifiers

CAS 1501-84-4
Molecular Formula C12H22ClN
Molecular Weight (g/mol) 215.77
MDL Number MFCD00072023
InChI Key OZBDFBJXRJWNAV-UHFFFAOYNA-N
Synonym rimantadine hydrochloride, flumadine, rimantadine hcl, meradane, 1-1-adamantyl ethylamine hydrochloride, 1-1-aminoethyl adamantane hydrochloride, alpha-methyl-1-adamantanemethylamine hydrochloride, 1-adamantan-1-yl ethanamine hydrochloride, algirem, meradan
PubChem CID 15165
ChEBI CHEBI:8865
IUPAC Name 1-(1-adamantyl)ethanamine;hydrochloride
SMILES [H+].[Cl-].CC(N)C12CC3CC(CC(C3)C1)C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BTB12981DA
SDS View Documents
Thermo Scientific Chemicals
BTB12981DA
1 g Amber Glass Bottle
Each for $19.14
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1501-84-4
215.77
OZBDFBJXRJWNAV-UHFFFAOYNA-N
15165
1-(1-adamantyl)ethanamine;hydrochloride
C12H22ClN
MFCD00072023
rimantadine hydrochloride, flumadine, rimantadine hcl, meradane, 1-1-adamantyl ethylamine hydrochloride, 1-1-aminoethyl adamantane hydrochloride, alpha-methyl-1-adamantanemethylamine hydrochloride, 1-adamantan-1-yl ethanamine hydrochloride, algirem, meradan
CHEBI:8865
[H+].[Cl-].CC(N)C12CC3CC(CC(C3)C1)C2

Specifications

1501-84-4
100.0
Amber Glass Bottle
MFCD00072023
rimantadine hydrochloride, flumadine, rimantadine hcl, meradane, 1-1-adamantyl ethylamine hydrochloride, 1-1-aminoethyl adamantane hydrochloride, alpha-methyl-1-adamantanemethylamine hydrochloride, 1-adamantan-1-yl ethanamine hydrochloride, algirem, meradan
[H+].[Cl-].CC(N)C12CC3CC(CC(C3)C1)C2
215.77
CHEBI:8865
95%
94.5
95%
C12H22ClN
1 g
OZBDFBJXRJWNAV-UHFFFAOYNA-N
1-(1-adamantyl)ethanamine;hydrochloride
15165
215.77
1-(1-Adamantyl)ethylamine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'