missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,4,4-Tetraphenyl-1,3-butadiene 99.0+%, TCI America™
Supplier: TCI America T01685G
Specifications
| 1,1,4,4-Tetraphenyl-1,3-butadiene | |
| 201°C | |
| C28H22 | |
| 5 g | |
| KLCLIOISYBHYDZ-UHFFFAOYSA-N | |
| (1,4,4-triphenylbuta-1,3-dien-1-yl)benzene | |
| 74060 | |
| ≥99.0% (HPLC) |
| 1450-63-1 | |
| White | |
| MFCD00004766 | |
| 1,1,4,4-tetraphenyl-1,3-butadiene, tetraphenylbutadiene, 1,1,4,4-tetraphenylbutadiene, 1,1,4,4-tetraphenylbuta-1,3-diene, 1,3-butadiene, 1,1,4,4-tetraphenyl, benzene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrakis, tetraphenyl butadiene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrabenzene, 1,4,4-triphenyl-1,3-butadienyl benzene, 1,4,4-tetraphenylbutadiene | |
| C(C=C(C1=CC=CC=C1)C1=CC=CC=C1)=C(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 358.48 | |
| 358.48 | |
| Crystalline Powder |
Chemical Identifiers
| 1450-63-1 | |
| 358.48 | |
| KLCLIOISYBHYDZ-UHFFFAOYSA-N | |
| 74060 | |
| C(C=C(C1=CC=CC=C1)C1=CC=CC=C1)=C(C1=CC=CC=C1)C1=CC=CC=C1 |
| C28H22 | |
| MFCD00004766 | |
| 1,1,4,4-tetraphenyl-1,3-butadiene, tetraphenylbutadiene, 1,1,4,4-tetraphenylbutadiene, 1,1,4,4-tetraphenylbuta-1,3-diene, 1,3-butadiene, 1,1,4,4-tetraphenyl, benzene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrakis, tetraphenyl butadiene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrabenzene, 1,4,4-triphenyl-1,3-butadienyl benzene, 1,4,4-tetraphenylbutadiene | |
| (1,4,4-triphenylbuta-1,3-dien-1-yl)benzene |
Safety and Handling
RTECSNumber : CY9040630
TSCA : Yes