missing translation for 'onlineSavingsMsg'
Learn More

1,1,4,4-Tetraphenyl-1,3-butadiene 99.0+%, TCI America™

Catalog No. T01681G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T01681G 1 g
T01685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T01681G Supplier TCI America Supplier No. T01681G

Chemical Identifiers

CAS 1450-63-1
Molecular Formula C28H22
Molecular Weight (g/mol) 358.48
MDL Number MFCD00004766
InChI Key KLCLIOISYBHYDZ-UHFFFAOYSA-N
Synonym 1,1,4,4-tetraphenyl-1,3-butadiene, tetraphenylbutadiene, 1,1,4,4-tetraphenylbutadiene, 1,1,4,4-tetraphenylbuta-1,3-diene, 1,3-butadiene, 1,1,4,4-tetraphenyl, benzene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrakis, tetraphenyl butadiene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrabenzene, 1,4,4-triphenyl-1,3-butadienyl benzene, 1,4,4-tetraphenylbutadiene
PubChem CID 74060
IUPAC Name (1,4,4-triphenylbuta-1,3-dien-1-yl)benzene
SMILES C(C=C(C1=CC=CC=C1)C1=CC=CC=C1)=C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 201°C
Color White
Quantity 1 g
Formula Weight 358.48
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1,4,4-Tetraphenyl-1,3-butadiene
RTECSNumber CY9040630
TSCA Yes