missing translation for 'onlineSavingsMsg'
Learn More

1,1,4,4-Tetraphenyl-1,3-butadiene, 99%

Catalog No. AAA1722503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1722503 1 g
AAA1722506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1722503 Supplier Thermo Scientific Chemicals Supplier No. A1722503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1450-63-1 | C28H22 | 358.48 g/mol

1,1,4,4-Tetraphenyl-1,3-butadiene is used as a wavelength shifter. It is also used as an electroluminescent dye, which glows blue with an electroluminescent dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,4,4-Tetraphenyl-1,3-butadiene is used as a wavelength shifter. It is also used as an electroluminescent dye, which glows blue with an electroluminescent dye.

Solubility
Soluble in ethanol, benzene, chloroform, tolune and acetic acid.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 1450-63-1
Molecular Formula C28H22
Molecular Weight (g/mol) 358.48
MDL Number MFCD00004766
InChI Key KLCLIOISYBHYDZ-UHFFFAOYSA-N
Synonym 1,1,4,4-tetraphenyl-1,3-butadiene, tetraphenylbutadiene, 1,1,4,4-tetraphenylbutadiene, 1,1,4,4-tetraphenylbuta-1,3-diene, 1,3-butadiene, 1,1,4,4-tetraphenyl, benzene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrakis, tetraphenyl butadiene, 1,1',1,1'-1,3-butadiene-1,4-diylidene tetrabenzene, 1,4,4-triphenyl-1,3-butadienyl benzene, 1,4,4-tetraphenylbutadiene
PubChem CID 74060
SMILES C(C=C(C1=CC=CC=C1)C1=CC=CC=C1)=C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 197°C to 202°C
Quantity 1 g
Solubility Information Soluble in ethanol,benzene,chloroform,tolune and acetic acid.
IUPAC Name 1,4,4-triphenylbuta-1,3-dienylbenzene
Formula Weight 358.48
Percent Purity 99%
Chemical Name or Material 1,1,4,4-Tetraphenyl-1,3-butadiene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 215-914-7
RTECSNumber CY9040630
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only