missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetramethylguanidine 99.0+%, TCI America™

Catalog No. T014825ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T014825ML 25 mL
T0148100ML 100 mL
T0148500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T014825ML Supplier TCI America Supplier No. T014825ML

Chemical Identifiers

CAS 80-70-6
Molecular Formula C5H13N3
Molecular Weight (g/mol) 115.18
MDL Number MFCD00008323
InChI Key KYVBNYUBXIEUFW-UHFFFAOYSA-N
Synonym tetramethylguanidine, n,n,n',n'-tetramethylguanidine, guanidine, n,n,n',n'-tetramethyl, tetramethyl guanidine, unii-vez101e7zu, 1,1,3,3-tetramethyl guanidine, n,n-1,1,3,3-tetramethylguanidine, vez101e7zu, 1,1,3,3-tetramethyl-guanidine, n,n,n',n'-tetramethyl-guanidine
PubChem CID 66460
IUPAC Name 1,1,3,3-tetramethylguanidine
SMILES CN(C)C(=N)N(C)C

Specifications

Melting Point -30°C
Color Colorless
Boiling Point 163°C
Quantity 25 mL
UN Number 2920
Formula Weight 115.18
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1,1,3,3-Tetramethylguanidine
EINECSNumber (2)-1774
TSCA Yes