accessibility menu, dialog, popup

UserName

CH3NH3CNHNCH3H3C

CAS RN 80-70-6

IUPAC Name: N,N,N',N'-tetramethylguanidine
Synonyms: 1,1,3,3-tetramethylguanidine
Molecular Weight (g/mol): 115.18
Molecular Formula: C5H13N3
InChi Key: KYVBNYUBXIEUFW-UHFFFAOYSA-N
SMILES: CN(C)C(=N)N(C)C
Boiling Point 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (5)