missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetramethylguanidine, 99%

Catalog No. p-7082059
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA12314AC 25 mL
AAA12314AE 100 mL
AAA12314AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA12314AC Supplier Thermo Scientific Chemicals Supplier No. A12314AC
Only null left

CAS: 80-70-6 | C5H13N3 | 115.18 g/mol

1,1,3,3-Tetramethylguanidine is employed as a polyurethane foam catalyst, as an accelerator for the syntheses of polysulfured rubber. It is also used as a strong base in the photochemical (couplers) and in the pharmaceutical (steroids) industries. It is essential for the preparation of alkyl nitriles from alkyl halides and 3'-alkylthymidines from 3'-nitrothymidines. It serves as an efficient and selective catalyst for the benzoylation of alcohols. It replaces the expensive bases 1,8-Diazabicyclo[5.4.0]undec-7-ene (DBU) and 1,5-Diazabicyclo[4.3.0]non-5-ene (DBN) in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,3,3-Tetramethylguanidine is employed as a polyurethane foam catalyst, as an accelerator for the syntheses of polysulfured rubber. It is also used as a strong base in the photochemical (couplers) and in the pharmaceutical (steroids) industries. It is essential for the preparation of alkyl nitriles from alkyl halides and 3′-alkylthymidines from 3′-nitrothymidines. It serves as an efficient and selective catalyst for the benzoylation of alcohols. It replaces the expensive bases 1,8-Diazabicyclo[5.4.0]undec-7-ene (DBU) and 1,5-Diazabicyclo[4.3.0]non-5-ene (DBN) in organic synthesis.

Solubility
Miscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents, mineral and organic acids, carbon dioxide. Keep away from sources of ignition.

Chemical Identifiers

CAS 80-70-6
Molecular Formula C5H13N3
Molecular Weight (g/mol) 115.18
MDL Number MFCD00008323
InChI Key KYVBNYUBXIEUFW-UHFFFAOYSA-N
Synonym tetramethylguanidine, n,n,n',n'-tetramethylguanidine, guanidine, n,n,n',n'-tetramethyl, tetramethyl guanidine, unii-vez101e7zu, 1,1,3,3-tetramethyl guanidine, n,n-1,1,3,3-tetramethylguanidine, vez101e7zu, 1,1,3,3-tetramethyl-guanidine, n,n,n',n'-tetramethyl-guanidine
PubChem CID 66460
IUPAC Name 1,1,3,3-tetramethylguanidine
SMILES CN(C)C(=N)N(C)C

Specifications

Melting Point <-30°C
Density 0.918
Boiling Point 162°C
Flash Point 60°C (140°F)
Refractive Index 1.469
Quantity 25 mL
UN Number UN2920
Beilstein 969608
Sensitivity Air Sensitive
Solubility Information Miscible with water.
Formula Weight 115.18
Percent Purity 99%
Chemical Name or Material 1,1,3,3-Tetramethylguanidine
Show More Show Less
Hazard Category H226-H302+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P331-P363-P372-P374-P380-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 201-302-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only