missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetramethylbutyl Isocyanide 95.0+%, TCI America™

Catalog No. T10541ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10541ML 1 mL
T10545ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T10541ML Supplier TCI America Supplier No. T10541ML

Chemical Identifiers

CAS 14542-93-9
Molecular Formula C9H17N
Molecular Weight (g/mol) 139.242
MDL Number MFCD00000003
InChI Key YVPXQMYCTGCWBE-UHFFFAOYSA-N
PubChem CID 26725
IUPAC Name 2-isocyano-2,4,4-trimethylpentane
SMILES CC(C)(C)CC(C)(C)[N+]#[C-]

Specifications

Color Colorless
Boiling Point 50°C
Quantity 1 mL
UN Number 1993
Formula Weight 139.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,3,3-Tetramethylbutyl Isocyanide
TSCA Yes
Recommended Storage Refrigerator