missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetramethyl-1,3-diphenyldisiloxane 98.0+%, TCI America™

Catalog No. t33485g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T33485G Supplier TCI America Supplier No. T33485G

Chemical Identifiers

CAS 56-33-7
Molecular Formula C16H22OSi2
Molecular Weight (g/mol) 286.52
MDL Number MFCD00039793
InChI Key YQJPWWLJDNCSCN-UHFFFAOYSA-N
PubChem CID 5945
ChEBI CHEBI:34061
IUPAC Name {[dimethyl(phenyl)silyl]oxy}dimethylphenylsilane
SMILES C[Si](C)(O[Si](C)(C)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 292°C
Quantity 5 g
Formula Weight 286.52
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,3,3-Tetramethyl-1,3-diphenyldisiloxane
EINECSNumber (7)-0474
RTECSNumber JM9236000
TSCA Yes