accessibility menu, dialog, popup

UserName

CH3SiH3COSiCH3H3C

CAS RN 56-33-7

IUPAC Name: {[dimethyl(phenyl)silyl]oxy}dimethylphenylsilane
Synonyms: bis(dimethylphenylsilyl)ether bis(phenyldimethylsilyl)oxide bis(dimethylphenylsilyl)oxide
Molecular Weight (g/mol): 286.52
Molecular Formula: C16H22OSi2
InChi Key: YQJPWWLJDNCSCN-UHFFFAOYSA-N
SMILES: C[Si](C)(O[Si](C)(C)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results