missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetramethoxypropane 98.0+%, TCI America™

Catalog No. M054425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M054425G 25 g
M0544500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M054425G Supplier TCI America Supplier No. M054425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 102-52-3
Molecular Formula C7H16O4
Molecular Weight (g/mol) 164.20
MDL Number MFCD00008488
InChI Key XHTYQFMRBQUCPX-UHFFFAOYSA-N
Synonym propane, 1,1,3,3-tetramethoxy, tetramethoxypropane, malonaldehyde bis dimethyl acetal, malonaldehyde tetramethyl acetal, malonaldehyde, bis dimethyl acetal, 1,1,3,3-tetramethoxy-propane, malonaldehyde bis dime, 1,3,3-tetramethoxypropane, ksc174o0b, propane,1,3,3-tetramethoxy
PubChem CID 66019
IUPAC Name 1,1,3,3-tetramethoxypropane
SMILES COC(CC(OC)OC)OC

Specifications

Color Yellow
Boiling Point 78°C
Quantity 25 g
Formula Weight 164.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,3,3-Tetramethoxypropane
EINECSNumber (2)-2459
TSCA Yes