missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetraethylurea, 99%

Catalog No. p-7045111
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0945806 5 g
AAL0945814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0945806 Supplier Thermo Scientific Chemicals Supplier No. L0945806
Only null left

CAS: 1187-03-7 | C9H20N2O | 172.272 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1187-03-7
Molecular Formula C9H20N2O
Molecular Weight (g/mol) 172.272
MDL Number MFCD00042881
InChI Key UWHSPZZUAYSGTB-UHFFFAOYSA-N
Synonym tetraethylurea, urea, tetraethyl, n,n,n',n'-tetraethylurea, unii-w511x6rp5r, urea, 1,1,3,3-tetraethyl, urea, n,n,n',n'-tetraethyl, teu, tetraethyl-urea, urea,tetraethyl, 1,1,3,3-tetraethyl-ure
PubChem CID 14465
IUPAC Name 1,1,3,3-tetraethylurea
SMILES CCN(CC)C(=O)N(CC)CC

Specifications

Density 0.91
Boiling Point 212°C to 214°C
Flash Point 81°C (177°F)
Refractive Index 1.447
Quantity 5 g
Beilstein 1763126
Formula Weight 172.27
Percent Purity 99%
Chemical Name or Material 1,1,3,3-Tetraethylurea
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 214-696-0
RTECSNumber YU2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only