missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetraethoxypropane 95.0+%, TCI America™

Catalog No. p-7136075
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
T009325ML 25 mL
T0093100ML 100 mL
T0093500ML 500 mL
3 options

Catalog No. T009325ML

Supplier: TCI America T009325ML

Only null left

Chemical Identifiers

CAS 122-31-6
Molecular Formula C11H24O4
Molecular Weight (g/mol) 220.309
MDL Number MFCD00009240
InChI Key KVJHGPAAOUGYJX-UHFFFAOYSA-N
Synonym malonaldehyde bis diethyl acetal, tetraethoxypropane, malonaldehyde tetraethyl acetal, malonaldehyde diethyl acetal, usaf kf-26, propane, 1,1,3,3-tetraethoxy, tetraethyl malondialdehyde acetal, tetraethoxy propane, malonaldehyde tetraethyl diacetal, malonaldehyde, bis diethyl acetal
PubChem CID 67147
IUPAC Name 1,1,3,3-tetraethoxypropane
SMILES CCOC(CC(OCC)OCC)OCC

Specifications

Color Colorless
Boiling Point 220°C
Quantity 25 mL
UN Number 3271
Formula Weight 220.31
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,3,3-Tetraethoxypropane
EINECSNumber (2)-2459
RTECSNumber ON8750000
TSCA No