accessibility menu, dialog, popup

UserName

CH3OOH3COH3COH3C

CAS RN 122-31-6

IUPAC Name: 1,1,3,3-tetraethoxypropane
Synonyms: 1,1,3,3-tetraethoxypropane
Molecular Weight (g/mol): 220.31
Molecular Formula: C11H24O4
InChi Key: KVJHGPAAOUGYJX-UHFFFAOYSA-N
SMILES: CCOC(CC(OCC)OCC)OCC
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (2)