missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,3,3-Tetraethoxypropane 95.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 220°C |
| Quantity | 500 mL |
| UN Number | 3271 |
| Formula Weight | 220.31 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1,1,3,3-Tetraethoxypropane |
Safety and Handling
| EINECSNumber | (2)-2459 |
| RTECSNumber | ON8750000 |
| TSCA | No |