missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetraethoxypropane 95.0+%, TCI America™

Catalog No. T009325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T009325ML 25 mL
T0093100ML 100 mL
T0093500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T009325ML Supplier TCI America Supplier No. T009325ML
Only null left

Chemical Identifiers

CAS 122-31-6
Molecular Formula C11H24O4
Molecular Weight (g/mol) 220.309
MDL Number MFCD00009240
InChI Key KVJHGPAAOUGYJX-UHFFFAOYSA-N
Synonym malonaldehyde bis diethyl acetal, tetraethoxypropane, malonaldehyde tetraethyl acetal, malonaldehyde diethyl acetal, usaf kf-26, propane, 1,1,3,3-tetraethoxy, tetraethyl malondialdehyde acetal, tetraethoxy propane, malonaldehyde tetraethyl diacetal, malonaldehyde, bis diethyl acetal
PubChem CID 67147
IUPAC Name 1,1,3,3-tetraethoxypropane
SMILES CCOC(CC(OCC)OCC)OCC

Specifications

Color Colorless
Boiling Point 220°C
Quantity 25 mL
UN Number 3271
Formula Weight 220.31
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,3,3-Tetraethoxypropane
EINECSNumber (2)-2459
RTECSNumber ON8750000
TSCA No