missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3,5,5-Hexamethyltrisiloxane 97.0+%, TCI America™

Catalog No. H14415ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14415ML 5 mL
H144125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14415ML Supplier TCI America Supplier No. H14415ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1189-93-1
Molecular Formula C6H18O2Si3
Molecular Weight (g/mol) 206.463
MDL Number MFCD00039788
InChI Key YTEISYFNYGDBRV-UHFFFAOYSA-N
Synonym 1,1,3,3,5,5-hexamethyltrisiloxane, trisiloxane, 1,1,3,3,5,5-hexamethyl, 1,3,3,5,5-hexamethyltrisiloxane, 2,4,4,6-tetramethyl-3,5-dioxa-2,4,6-trisilaheptane, dimethylbis dimethylsilyloxy silane
PubChem CID 6327152
IUPAC Name [(dimethyl-$l^{3}-silanyl)oxy-dimethylsilyl]oxy-dimethylsilicon
SMILES C[Si](C)O[Si](C)(C)O[Si](C)C

Specifications

Boiling Point 128°C
Color Colorless
Physical Form Liquid
Quantity 5 mL
UN Number 1993
Formula Weight 208.48
Percent Purity ≥97.0% (GC)
Chemical Name or Material 1,1,3,3,5,5-Hexamethyltrisiloxane
EINECSNumber (7)-0477
TSCA Yes