accessibility menu, dialog, popup

UserName

H3CSiCH3OSiH3CCH3OSiCH3H3C

CAS RN 1189-93-1

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 206.46
Molecular Formula: C6H18O2Si3
InChi Key: YTEISYFNYGDBRV-UHFFFAOYSA-N
SMILES: C[Si](C)O[Si](C)(C)O[Si](C)C
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)