missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3,5,5-Hexamethyltrisiloxane, 95%

Catalog No. p-7046837
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1652022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1652022 Supplier Thermo Scientific Chemicals Supplier No. L1652022

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1189-93-1 | C6H18O2Si3 | 206.463 g/mol

1,1,3,3,5,5-Hexamethyltrisiloxane is involved in the preparation of 2-bis(3-allyl-4-hydroxyphenyl)hexafluoropropane, which is associated with inorganic siloxane polymer and used as a coating material for the detection of toxic chemical warfare agents. It is also used as a silylating agent and to prepare 1,5-Bis[2-(1,2-epoxycyclohex-4-yl)ethyl]-1,1,3,3,5,5-hexamethyltrisiloxane from 4-vinyl-1-cyclohexene-1,2-epoxide by hydrosilylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,3,3,5,5-Hexamethyltrisiloxane is involved in the preparation of 2-bis(3-allyl-4-hydroxyphenyl)hexafluoropropane, which is associated with inorganic siloxane polymer and used as a coating material for the detection of toxic chemical warfare agents. It is also used as a silylating agent and to prepare 1,5-Bis[2-(1,2-epoxycyclohex-4-yl)ethyl]-1,1,3,3,5,5-hexamethyltrisiloxane from 4-vinyl-1-cyclohexene-1,2-epoxide by hydrosilylation.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 1189-93-1
Molecular Formula C6H18O2Si3
Molecular Weight (g/mol) 206.463
MDL Number MFCD00039788
InChI Key YTEISYFNYGDBRV-UHFFFAOYSA-N
Synonym 1,1,3,3,5,5-hexamethyltrisiloxane, trisiloxane, 1,1,3,3,5,5-hexamethyl, 1,3,3,5,5-hexamethyltrisiloxane, 2,4,4,6-tetramethyl-3,5-dioxa-2,4,6-trisilaheptane, dimethylbis dimethylsilyloxy silane
PubChem CID 6327152
IUPAC Name [(dimethyl-$l^{3}-silanyl)oxy-dimethylsilyl]oxy-dimethylsilicon
SMILES C[Si](C)O[Si](C)(C)O[Si](C)C

Specifications

Boiling Point 128°C
Melting Point -67°C
Quantity 100 g
Flash Point 20°C (68°F)
Refractive Index 1.381
Sensitivity Moisture sensitive
Density 0.822
UN Number UN1993
Beilstein 1746341
Solubility Information Slightly miscible with water.
Formula Weight 208.48
Percent Purity 97%
Chemical Name or Material 1,1,3,3,5,5-Hexamethyltrisiloxane
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 214-716-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only