missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3,5,5-Hexaethoxy-1,3,5-trisilacyclohexane 90.0+%, TCI America™

Catalog No. h15131g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H15131G Supplier TCI America Supplier No. H15131G

Chemical Identifiers

CAS 17955-67-8
Molecular Formula C15H36O6Si3
Molecular Weight (g/mol) 396.70
MDL Number MFCD06660097
InChI Key SPEDTQCGQOZHQP-UHFFFAOYSA-N
Synonym 1,1,3,3,5,5-Hexaethoxy-1,3,5-trisilinane
PubChem CID 10960278
IUPAC Name 1,1,3,3,5,5-hexaethoxy-1,3,5-trisilinane
SMILES CCO[Si]1(C[Si](C[Si](C1)(OCC)OCC)(OCC)OCC)OCC

Specifications

Color Colorless
Boiling Point 133°C
Quantity 1 g
Formula Weight 396.70
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,3,3,5,5-Hexaethoxy-1,3,5-trisilacyclohexane