accessibility menu, dialog, popup

UserName

H3COSiSiSiOCH3OH3COCH3OCH3OCH3

CAS RN 17955-67-8

IUPAC Name: 1,1,3,3,5,5-hexaethoxy-1,3,5-trisilinane
Synonyms: 1,1,3,3,5,5-hexaethoxy-1,3,5-trisilinane
Molecular Weight (g/mol): 396.7
Molecular Formula: C15H36O6Si3
InChi Key: SPEDTQCGQOZHQP-UHFFFAOYSA-N
SMILES: CCO[Si]1(C[Si](C[Si](C1)(OCC)OCC)(OCC)OCC)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results