missing translation for 'onlineSavingsMsg'
Learn More

1,1,2-Trimethyl-1H-benzo[e]indole 98.0+%, TCI America™

Catalog No. T17145G
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17145G 5 g
T171425G 25 g
T1714100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T17145G Supplier TCI America Supplier No. T17145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41532-84-7
Molecular Formula C15H15N
Molecular Weight (g/mol) 209.29
MDL Number MFCD00082627
InChI Key WJZSZXCWMATYFX-UHFFFAOYSA-N
Synonym 1,1,2-trimethyl-1h-benzo e indole, 1,1,2-trimethyl-1h-benz e indole, 1,1,2-trimethylbenz e indole, 2,3,3-trimethyl-4,5-benzo-3h-indole, 2,3,3-trimethylbenzoindolenine, 1,1,2-trimethylbenzo e indole, 1h-benz e indole, 1,1,2-trimethyl, 2,3,3-trimethyl-1h-benz e indolenine, 1,1,2-trimethylhydrobenzo e indole
PubChem CID 170530
IUPAC Name 1,1,2-trimethyl-1H-benzo[e]indole
SMILES CC1=NC2=CC=C3C=CC=CC3=C2C1(C)C

Specifications

Melting Point 115°C
Quantity 5 g
Formula Weight 209.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1,2-Trimethyl-1H-benzo[e]indole