accessibility menu, dialog, popup

UserName

H3CNCH3H3C

CAS RN 41532-84-7

IUPAC Name: 1,1,2-trimethyl-1H-benzo[e]indole
Synonyms: 1,1,2-trimethylbenzo[e]indole
Molecular Weight (g/mol): 209.29
Molecular Formula: C15H15N
InChi Key: WJZSZXCWMATYFX-UHFFFAOYSA-N
SMILES: CC1=NC2=CC=C3C=CC=CC3=C2C1(C)C
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)