missing translation for 'onlineSavingsMsg'
Learn More

1,1,2-Trimethyl-1H-benzo[e]indole, 97%

Catalog No. p-7026032
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2180406 5 g
AAB2180414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2180406 Supplier Thermo Scientific Chemicals Supplier No. B2180406

CAS: 41532-84-7 | C15H15N | 209.29 g/mol

Reactant used for preparation of fluorescent probes for imaging of tumors, hypoxia-sensitive fluorescent probes for real-time imaging of acute ischemia and near IR fluorescent probe for imaging integrin receptor expression.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant used for preparation of fluorescent probes for imaging of tumors , hypoxia-sensitive fluorescent probes for real-time imaging of acute ischemia and near IR fluorescent probe for imaging integrin receptor expression.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 41532-84-7
Molecular Formula C15H15N
Molecular Weight (g/mol) 209.29
MDL Number MFCD00082627
InChI Key WJZSZXCWMATYFX-UHFFFAOYSA-N
Synonym 1,1,2-trimethyl-1h-benzo e indole, 1,1,2-trimethyl-1h-benz e indole, 1,1,2-trimethylbenz e indole, 2,3,3-trimethyl-4,5-benzo-3h-indole, 2,3,3-trimethylbenzoindolenine, 1,1,2-trimethylbenzo e indole, 1h-benz e indole, 1,1,2-trimethyl, 2,3,3-trimethyl-1h-benz e indolenine, 1,1,2-trimethylhydrobenzo e indole
PubChem CID 170530
IUPAC Name 1,1,2-trimethylbenzo[e]indole
SMILES CC1=NC2=CC=C3C=CC=CC3=C2C1(C)C

Specifications

Melting Point 114°C to 119°C
Quantity 5 g
Beilstein 153709
Solubility Information Slightly soluble in water.
Formula Weight 209.29
Percent Purity 97%
Chemical Name or Material 1,1,2-Trimethyl-1H-benzo[e]indole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 255-429-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only