missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,2-Trimethoxyethane 98.0+%, TCI America™
Supplier: TCI America T0831100ML
Specifications
| 1,1,2-Trimethoxyethane | |
| Colorless | |
| C5H12O3 | |
| 100 mL | |
| Methoxyacetaldehyde Dimethyl Acetal | |
| COCC(OC)OC | |
| 120.15 | |
| 120.15 | |
| Liquid |
| 24332-20-5 | |
| 126°C | |
| MFCD00008486 | |
| 3271 | |
| DYOZNCVZPFIXLU-UHFFFAOYSA-N | |
| 1,1,2-trimethoxyethane | |
| 47520 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 24332-20-5 | |
| 120.15 | |
| DYOZNCVZPFIXLU-UHFFFAOYSA-N | |
| 47520 | |
| COCC(OC)OC |
| C5H12O3 | |
| MFCD00008486 | |
| Methoxyacetaldehyde Dimethyl Acetal | |
| 1,1,2-trimethoxyethane |
Safety and Handling
TSCA : No