missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,2-Trimethoxyethane 98.0+%, TCI America™
$311.30 - $932.03
Chemical Identifiers
| CAS | 24332-20-5 |
|---|---|
| Molecular Formula | C5H12O3 |
| Molecular Weight (g/mol) | 120.15 |
| MDL Number | MFCD00008486 |
| InChI Key | DYOZNCVZPFIXLU-UHFFFAOYSA-N |
| Synonym | Methoxyacetaldehyde Dimethyl Acetal |
| PubChem CID | 47520 |
| IUPAC Name | 1,1,2-trimethoxyethane |
| SMILES | COCC(OC)OC |
Chemical Identifiers
| 24332-20-5 | |
| 120.15 | |
| DYOZNCVZPFIXLU-UHFFFAOYSA-N | |
| 47520 | |
| COCC(OC)OC |
| C5H12O3 | |
| MFCD00008486 | |
| Methoxyacetaldehyde Dimethyl Acetal | |
| 1,1,2-trimethoxyethane |
Specifications
| 24332-20-5 | |
| 126°C | |
| MFCD00008486 | |
| 3271 | |
| DYOZNCVZPFIXLU-UHFFFAOYSA-N | |
| 1,1,2-trimethoxyethane | |
| 47520 | |
| ≥98.0% (GC) | |
| 1,1,2-Trimethoxyethane |
| Colorless | |
| C5H12O3 | |
| 25 mL | |
| Methoxyacetaldehyde Dimethyl Acetal | |
| COCC(OC)OC | |
| 120.15 | |
| 120.15 | |
| Liquid |
Safety and Handling
TSCA : No