missing translation for 'onlineSavingsMsg'
Learn More

1,1,2-Trichloroethane 98.0+%, TCI America™

Catalog No. T038125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T038125G 25 g
T0381500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T038125G Supplier TCI America Supplier No. T038125G

Chemical Identifiers

CAS 79-00-5
Molecular Formula C2H3Cl3
Molecular Weight (g/mol) 133.40
MDL Number MFCD00000852
InChI Key UBOXGVDOUJQMTN-UHFFFAOYSA-N
Synonym ethane, 1,1,2-trichloro, vinyltrichloride, beta-trichloroethane, 1,1,2-trichlorethane, vinyl trichloride, 1,2,2-trichloroethane, ethane trichloride, rcra waste number u227, rcra waste number u359, .beta.-t
PubChem CID 6574
ChEBI CHEBI:36018
IUPAC Name 1,1,2-trichloroethane
SMILES ClCC(Cl)Cl

Specifications

Color Colorless
Boiling Point 114°C
Quantity 25 g
UN Number 2810
Formula Weight 133.40
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,2-Trichloroethane
EINECSNumber (2)-0055
RTECSNumber KJ3150000
TSCA Yes