accessibility menu, dialog, popup

UserName

ClClCl

CAS RN 79-00-5

IUPAC Name: 1,1,2-trichloroethane
Synonyms: 1,1, 2-trichloroethane β-trichloroethane epa pesticide chemical code 081203 trojchloroetan(1,1,2) ccris 602 rcra waste number U227 rcra waste number U359 caswell no. 875A 1,1,2-trichloraethan
Molecular Weight (g/mol): 133.4
Molecular Formula: C2H3Cl3
InChi Key: UBOXGVDOUJQMTN-UHFFFAOYSA-N
SMILES: ClCC(Cl)Cl
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)