missing translation for 'onlineSavingsMsg'
Learn More

1,1,2-Tribromoethane 99.0+%, TCI America™

Catalog No. t077325ml
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T077325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T077325ML Supplier TCI America Supplier No. T077325ML

Chemical Identifiers

CAS 78-74-0
Molecular Formula C2H3Br3
Molecular Weight (g/mol) 266.758
MDL Number MFCD00039168
InChI Key QUMDOMSJJIFTCA-UHFFFAOYSA-N
PubChem CID 66222
IUPAC Name 1,1,2-tribromoethane
SMILES C(C(Br)Br)Br

Specifications

Color Yellow
Boiling Point 188°C
Quantity 25 mL
Formula Weight 266.76
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,2-Tribromoethane