missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,3-Tetramethyl-1H-benzo[e]indolium Hexafluorophosphate 98.0+%, TCI America™

Catalog No. T34575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34575G 5 g
T345725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T34575G Supplier TCI America Supplier No. T34575G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 372081-65-7
Molecular Formula C16H18F6NP
Molecular Weight (g/mol) 369.29
MDL Number MFCD16621101
InChI Key OBEJDMPKHNFDIA-UHFFFAOYSA-N
PubChem CID 11199491
IUPAC Name 1,1,2,3-tetramethyl-1H-benzo[e]indol-3-ium; hexafluoro-λ⁵-phosphanuide
SMILES F[P-](F)(F)(F)(F)F.CC1=[N+](C)C2=CC=C3C=CC=CC3=C2C1(C)C

Specifications

Color Purple
Quantity 5 g
Formula Weight 369.29
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,1,2,3-Tetramethyl-1H-benzo[e]indolium Hexafluorophosphate