missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,3,4,5-Hexaphenylsilole 98.0+%, TCI America™

Catalog No. H14131G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H14131G Supplier TCI America Supplier No. H14131G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 752-28-3
Molecular Formula C40H30Si
Molecular Weight (g/mol) 538.77
MDL Number MFCD27976827
InChI Key QAKMXYFDVPDIPT-UHFFFAOYSA-N
Synonym 1,1,2,3,4,5-Hexaphenylsilacyclopenta-2,4-diene
PubChem CID 20758316
IUPAC Name hexaphenyl-1H-silole
SMILES C1=CC=C(C=C1)C1=C(C2=CC=CC=C2)[Si](C(=C1C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 195°C
Quantity 1 g
Formula Weight 538.77
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1,2,3,4,5-Hexaphenylsilole