accessibility menu, dialog, popup

UserName

Si

CAS RN 752-28-3

IUPAC Name: hexaphenyl-1H-silole
Synonyms: hexaphenylsilole
Molecular Weight (g/mol): 538.77
Molecular Formula: C40H30Si
InChi Key: QAKMXYFDVPDIPT-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=C(C2=CC=CC=C2)[Si](C(=C1C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results