missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,3,3,3-Hexafluoropropyl Methyl Ether 98.0+%, TCI America™

Catalog No. H15075G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15075G 5 g
H150725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15075G Supplier TCI America Supplier No. H15075G

Chemical Identifiers

CAS 382-34-3
Molecular Formula C4H4F6O
Molecular Weight (g/mol) 182.065
MDL Number MFCD00041537
InChI Key PKMXTDVNDDDCSY-UHFFFAOYSA-N
PubChem CID 2778364
IUPAC Name 1,1,1,2,3,3-hexafluoro-3-methoxypropane
SMILES COC(C(C(F)(F)F)F)(F)F

Specifications

Color Colorless
Boiling Point 55°C
Quantity 5 g
UN Number 3271
Formula Weight 182.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,2,3,3,3-Hexafluoropropyl Methyl Ether
EINECSNumber (2)-3998
TSCA No