accessibility menu, dialog, popup

UserName

CH3OFFFFFF

CAS RN 382-34-3

IUPAC Name: 1,1,1,2,3,3-hexafluoro-3-methoxypropane
Synonyms: 1,1,1,2,3,3-hexafluoro-3-methoxypropane
Molecular Weight (g/mol): 182.07
Molecular Formula: C4H4F6O
InChi Key: PKMXTDVNDDDCSY-UHFFFAOYNA-N
SMILES: COC(F)(F)C(F)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results