missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetramethylcyclopropane 98.0+%, TCI America™

Catalog No. T17215ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17215ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T17215ML Supplier TCI America Supplier No. T17215ML

Chemical Identifiers

CAS 4127-47-3
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00060790
InChI Key JCHUCGKEGUAHEH-UHFFFAOYSA-N
Synonym cyclopropane, 1,1,2,2-tetramethyl, acmc-209jjk, jchucgkeguaheh-uhfffaoysa, 1,1,2,2-tetramethyl cyclopropane, 1,1,2,2-tetramethyl-cyclopropane
PubChem CID 77778
IUPAC Name 1,1,2,2-tetramethylcyclopropane
SMILES CC1(C)CC1(C)C

Specifications

Color Colorless
Boiling Point 76°C
Quantity 5 mL
UN Number 3295
Formula Weight 98.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,2,2-Tetramethylcyclopropane