accessibility menu, dialog, popup

UserName

CH3H3CH3CH3C

CAS RN 4127-47-3

IUPAC Name: 1,1,2,2-tetramethylcyclopropane
Synonyms: 1,1,2,2-tetramethylcyclopropane
Molecular Weight (g/mol): 98.19
Molecular Formula: C7H14
InChi Key: JCHUCGKEGUAHEH-UHFFFAOYSA-N
SMILES: CC1(C)CC1(C)C
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results