missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetramethylcyclopropane, 99%, Thermo Scientific Chemicals

Catalog No. p-7025370
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2080906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2080906 Supplier Thermo Scientific Chemicals Supplier No. B2080906

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4127-47-3
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00060790
InChI Key JCHUCGKEGUAHEH-UHFFFAOYSA-N
Synonym cyclopropane, 1,1,2,2-tetramethyl, acmc-209jjk, jchucgkeguaheh-uhfffaoysa, 1,1,2,2-tetramethyl cyclopropane, 1,1,2,2-tetramethyl-cyclopropane
PubChem CID 77778
IUPAC Name 1,1,2,2-tetramethylcyclopropane
SMILES CC1(C)CC1(C)C

Specifications

Density 0.72
Boiling Point 76°C
Quantity 5 g
UN Number UN3295
Formula Weight 98.19
Percent Purity 99%
Chemical Name or Material 1,1,2,2-Tetramethylcyclopropane
Hazard Category H225-H304
Hazard Statement GHS H Statement
H224-H304
Extremely flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 223-937-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only