missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,2,2-Tetrafluoroethyl 2,2,3,3-Tetrafluoropropyl Ether 95%, TCI America™
$44.28 - $109.74
Chemical Identifiers
| CAS | 16627-68-2 |
|---|---|
| Molecular Formula | C5H4F8O |
| Molecular Weight (g/mol) | 232.07 |
| MDL Number | MFCD00155961 |
| InChI Key | HCBRSIIGBBDDCD-UHFFFAOYSA-N |
| PubChem CID | 2776662 |
| IUPAC Name | 1,1,2,2-tetrafluoro-3-(1,1,2,2-tetrafluoroethoxy)propane |
| SMILES | FC(F)C(F)(F)COC(F)(F)C(F)F |
Chemical Identifiers
| 16627-68-2 | |
| 232.07 | |
| HCBRSIIGBBDDCD-UHFFFAOYSA-N | |
| 1,1,2,2-tetrafluoro-3-(1,1,2,2-tetrafluoroethoxy)propane |
| C5H4F8O | |
| MFCD00155961 | |
| 2776662 | |
| FC(F)C(F)(F)COC(F)(F)C(F)F |
Specifications
| 16627-68-2 | |
| 92°C | |
| MFCD00155961 | |
| 3271 | |
| FC(F)C(F)(F)COC(F)(F)C(F)F | |
| 232.07 | |
| 232.07 | |
| Liquid |
| Colorless | |
| C5H4F8O | |
| 5 g | |
| HCBRSIIGBBDDCD-UHFFFAOYSA-N | |
| 1,1,2,2-tetrafluoro-3-(1,1,2,2-tetrafluoroethoxy)propane | |
| 2776662 | |
| ≥95.0% (GC) | |
| 1,1,2,2-Tetrafluoroethyl 2,2,3,3-Tetrafluoropropyl Ether |
Safety and Handling
EINECSNumber : (2)-4074
TSCA : No