accessibility menu, dialog, popup

UserName

FFFFOFFFF

CAS RN 16627-68-2

IUPAC Name: 1,1,2,2-tetrafluoro-3-(1,1,2,2-tetrafluoroethoxy)propane
Synonyms: 1,1,2,2-tetrafluoro-3-(1,1,2,2-tetrafluoroethoxy)propane
Molecular Weight (g/mol): 232.07
Molecular Formula: C5H4F8O
InChi Key: HCBRSIIGBBDDCD-UHFFFAOYSA-N
SMILES: FC(F)C(F)(F)COC(F)(F)C(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results