missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,2,2-Tetrafluoroethyl 2,2,3,3-Tetrafluoropropyl Ether 95%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 92°C |
| Quantity | 5 g |
| UN Number | 3271 |
| Formula Weight | 232.07 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1,1,2,2-Tetrafluoroethyl 2,2,3,3-Tetrafluoropropyl Ether |
Safety and Handling
| EINECSNumber | (2)-4074 |
| TSCA | No |