missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2,3,3-Hexafluoro-1,3-bis[(1,2,2-trifluorovinyl)oxy]propane 98.0+%, TCI America™

Catalog No. H16115G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H16115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H16115G Supplier TCI America Supplier No. H16115G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13846-22-5
Molecular Formula C7F12O2
Molecular Weight (g/mol) 344.056
InChI Key KGJWCQOEERZJMB-UHFFFAOYSA-N
PubChem CID 83771
IUPAC Name 1,1,2,2,3,3-hexafluoro-1,3-bis(1,2,2-trifluoroethenoxy)propane
SMILES C(=C(F)F)(OC(C(C(OC(=C(F)F)F)(F)F)(F)F)(F)F)F

Specifications

Color Colorless
Quantity 5 g
Formula Weight 344.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,2,2,3,3-Hexafluoro-1,3-bis[(1,2,2-trifluorovinyl)oxy]propane
TSCA No
Recommended Storage Refrigerator