accessibility menu, dialog, popup

UserName

CAS RN 13846-22-5

FFFOFFFFFFOFFF

CAS RN 13846-22-5

IUPAC Name: 1,1,2,2,3,3-hexafluoro-1,3-bis[(1,2,2-trifluoroethenyl)oxy]propane
Synonyms: 1,1,2,2,3,3-hexafluoro-1,3-bis[(1,2,2-trifluoroethenyl)oxy]propane
Molecular Weight (g/mol): 344.06
Molecular Formula: C7F12O2
InChi Key: KGJWCQOEERZJMB-UHFFFAOYSA-N
SMILES: FC(F)=C(F)OC(F)(F)C(F)(F)C(F)(F)OC(F)=C(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results