missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,1-Tris(4-hydroxyphenyl)ethane 98.0+%, TCI America™
$80.16 - $598.37
Chemical Identifiers
| CAS | 27955-94-8 |
|---|---|
| Molecular Formula | C20H18O3 |
| Molecular Weight (g/mol) | 306.361 |
| MDL Number | MFCD00012180 |
| InChI Key | BRPSWMCDEYMRPE-UHFFFAOYSA-N |
| PubChem CID | 93118 |
| IUPAC Name | 4-[1,1-bis(4-hydroxyphenyl)ethyl]phenol |
| SMILES | CC(C1=CC=C(C=C1)O)(C2=CC=C(C=C2)O)C3=CC=C(C=C3)O |
Chemical Identifiers
| 27955-94-8 | |
| 306.361 | |
| BRPSWMCDEYMRPE-UHFFFAOYSA-N | |
| 4-[1,1-bis(4-hydroxyphenyl)ethyl]phenol |
| C20H18O3 | |
| MFCD00012180 | |
| 93118 | |
| CC(C1=CC=C(C=C1)O)(C2=CC=C(C=C2)O)C3=CC=C(C=C3)O |
Specifications
| 27955-94-8 | |
| White | |
| MFCD00012180 | |
| 3077 | |
| CC(C1=CC=C(C=C1)O)(C2=CC=C(C=C2)O)C3=CC=C(C=C3)O | |
| 306.361 | |
| 306.36 | |
| Crystalline Powder |
| 248°C | |
| C20H18O3 | |
| 25 g | |
| BRPSWMCDEYMRPE-UHFFFAOYSA-N | |
| 4-[1,1-bis(4-hydroxyphenyl)ethyl]phenol | |
| 93118 | |
| ≥98.0% (GC) | |
| 1,1,1-Tris(4-hydroxyphenyl)ethane |
Safety and Handling
EINECSNumber : (4)-1668
RTECSNumber : SL4375275
TSCA : Yes