missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Tris(4-hydroxyphenyl)ethane 98.0+%, TCI America™

Catalog No. T125425G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T125425G 25 g
T1254100G 100 g
T1254500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T125425G Supplier TCI America Supplier No. T125425G
Only null left

Chemical Identifiers

CAS 27955-94-8
Molecular Formula C20H18O3
Molecular Weight (g/mol) 306.361
MDL Number MFCD00012180
InChI Key BRPSWMCDEYMRPE-UHFFFAOYSA-N
PubChem CID 93118
IUPAC Name 4-[1,1-bis(4-hydroxyphenyl)ethyl]phenol
SMILES CC(C1=CC=C(C=C1)O)(C2=CC=C(C=C2)O)C3=CC=C(C=C3)O

Specifications

Melting Point 248°C
Color White
Quantity 25 g
UN Number 3077
Formula Weight 306.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1,1-Tris(4-hydroxyphenyl)ethane
EINECSNumber (4)-1668
RTECSNumber SL4375275
TSCA Yes