missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoro-5-methyl-2,4-hexanedione, 97%, Thermo Scientific™

Catalog No. AC421760250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC421760250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC421760250 Supplier Thermo Scientific Chemicals Supplier No. 421760250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 30984-28-2
Molecular Formula C7H9F3O2
Molecular Weight (g/mol) 182.14
MDL Number MFCD00045106
InChI Key QDJWKASBKAMIDF-ZZXKWVIFSA-N
Synonym 1,1,1-trifluoro-5-methyl-2,4-hexanedione, trifluoroacetyl isobutyrylmethane, 2,4-hexanedione, 1,1,1-trifluoro-5-methyl, 5-methyl-1,1,1-trifluoro-2,4-hexanedione, 2,4-hexanedione,1,1,1-trifluoro-5-methyl, tos-bb-0654, 2,4-dioxo-5-methyl-1,1,1-trifluorohexane, 5-methyl-1,1,1-trifluorohexane-2,4-dione
PubChem CID 160191
IUPAC Name 1,1,1-trifluoro-5-methylhexane-2,4-dione
SMILES CC(C)C(=O)\C=C(\O)C(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Boiling Point 61°C (48.0 mmHg)
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4005 to 1.4025
Linear Formula CF3COCH2COCH(CH3)2
Quantity 25 g
Formula Weight 182.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1,1,1-Trifluoro-5-methyl-2,4-hexanedione
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 250-415-8

RUO – Research Use Only