CAS RN 30984-28-2
IUPAC Name:
(4E)-6,6,6-trifluoro-5-hydroxy-2-methylhex-4-en-3-one
Synonyms:
(4E)-6,6,6-trifluoro-5-hydroxy-2-methylhex-4-en-3-one
Molecular Weight (g/mol):
182.14
Molecular Formula:
C7H9F3O2
InChi Key:
QDJWKASBKAMIDF-ZZXKWVIFSA-N
SMILES:
CC(C)C(=O)\C=C(\O)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
1,1,1-Trifluoro-5-methyl-2,4-hexanedione, 97%, Thermo Scientific™
1,1,1-Trifluoro-5-methyl-2,4-hexanedione, 97%, Thermo Scientific™
CAS: 30984-28-2 Molecular Formula: C7H9F3O2 Molecular Weight (g/mol): 182.14 MDL Number: MFCD00045106 InChI Key: QDJWKASBKAMIDF-ZZXKWVIFSA-N Synonym: 1,1,1-trifluoro-5-methyl-2,4-hexanedione,trifluoroacetyl isobutyrylmethane,2,4-hexanedione, 1,1,1-trifluoro-5-methyl,5-methyl-1,1,1-trifluoro-2,4-hexanedione,2,4-hexanedione,1,1,1-trifluoro-5-methyl,tos-bb-0654,2,4-dioxo-5-methyl-1,1,1-trifluorohexane,5-methyl-1,1,1-trifluorohexane-2,4-dione PubChem CID: 160191 IUPAC Name: 1,1,1-trifluoro-5-methylhexane-2,4-dione SMILES: CC(C)C(=O)\C=C(\O)C(F)(F)F