missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,1-Trifluoro-4-iodobutane 98.0+%, TCI America™
$148.25 - $433.16
Chemical Identifiers
| CAS | 461-17-6 |
|---|---|
| Molecular Formula | C4H6F3I |
| Molecular Weight (g/mol) | 237.992 |
| MDL Number | MFCD00041543 |
| InChI Key | LNDGACQEAYKNOI-UHFFFAOYSA-N |
| Synonym | 1-iodo-4,4,4-trifluorobutane, 4,4,4-trifluoro-1-iodobutane, 4,4,4-trifluorobutyl iodide, 4-iodo-1,1,1-trifluorobutane, butane, 1,1,1-trifluoro-4-iodo, 1,1,1-trifluoro-4-iodo-butane, acmc-209k5v, 4,4,4-trifluorobutyliodide, ksc235m2j, 1-iodo-4,4,4-trifluoro-butane |
| PubChem CID | 2775189 |
| IUPAC Name | 1,1,1-trifluoro-4-iodobutane |
| SMILES | C(CC(F)(F)F)CI |
Chemical Identifiers
| 461-17-6 | |
| 237.992 | |
| LNDGACQEAYKNOI-UHFFFAOYSA-N | |
| 2775189 | |
| C(CC(F)(F)F)CI |
| C4H6F3I | |
| MFCD00041543 | |
| 1-iodo-4,4,4-trifluorobutane, 4,4,4-trifluoro-1-iodobutane, 4,4,4-trifluorobutyl iodide, 4-iodo-1,1,1-trifluorobutane, butane, 1,1,1-trifluoro-4-iodo, 1,1,1-trifluoro-4-iodo-butane, acmc-209k5v, 4,4,4-trifluorobutyliodide, ksc235m2j, 1-iodo-4,4,4-trifluoro-butane | |
| 1,1,1-trifluoro-4-iodobutane |
Specifications
| 461-17-6 | |
| C4H6F3I | |
| 5 g | |
| LNDGACQEAYKNOI-UHFFFAOYSA-N | |
| 1,1,1-trifluoro-4-iodobutane | |
| 2775189 | |
| ≥98.0% (GC) | |
| 1,1,1-Trifluoro-4-iodobutane |
| 128°C | |
| MFCD00041543 | |
| 1-iodo-4,4,4-trifluorobutane, 4,4,4-trifluoro-1-iodobutane, 4,4,4-trifluorobutyl iodide, 4-iodo-1,1,1-trifluorobutane, butane, 1,1,1-trifluoro-4-iodo, 1,1,1-trifluoro-4-iodo-butane, acmc-209k5v, 4,4,4-trifluorobutyliodide, ksc235m2j, 1-iodo-4,4,4-trifluoro-butane | |
| C(CC(F)(F)F)CI | |
| 237.992 | |
| 237.99 | |
| Liquid |
Safety and Handling
TSCA : No