missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,1-Trifluoro-3-iodopropane 98.0+%, TCI America™
Supplier: TCI America T258825G
Specifications
| 1,1,1-Trifluoro-3-iodopropane | |
| Yellow | |
| C3H4F3I | |
| 25 g | |
| ULIYQAUQKZDZOX-UHFFFAOYSA-N | |
| 1,1,1-trifluoro-3-iodopropane | |
| 156165 | |
| ≥98.0% (GC) |
| 460-37-7 | |
| 90°C | |
| MFCD00038531 | |
| 1-iodo-3,3,3-trifluoropropane, 3,3,3-trifluoropropyl iodide, 3-iodo-1,1,1-trifluoropropane, 1-iodo-3,3,3-trifluoro propane, 3,3,3-trifluoropropyliodide, 1-iodo-2-perfluoroalkyl ethane, 1,1,1-trifluoro-3-iodo-propane, propane, 1,1,1-trifluoro-3-iodo, trifluoroiodopropane | |
| C(CI)C(F)(F)F | |
| 223.965 | |
| 223.96 | |
| Liquid |
Chemical Identifiers
| 460-37-7 | |
| 223.965 | |
| ULIYQAUQKZDZOX-UHFFFAOYSA-N | |
| 156165 | |
| C(CI)C(F)(F)F |
| C3H4F3I | |
| MFCD00038531 | |
| 1-iodo-3,3,3-trifluoropropane, 3,3,3-trifluoropropyl iodide, 3-iodo-1,1,1-trifluoropropane, 1-iodo-3,3,3-trifluoro propane, 3,3,3-trifluoropropyliodide, 1-iodo-2-perfluoroalkyl ethane, 1,1,1-trifluoro-3-iodo-propane, propane, 1,1,1-trifluoro-3-iodo, trifluoroiodopropane | |
| 1,1,1-trifluoro-3-iodopropane |
Safety and Handling
TSCA : No